Assay Details
| Gene Symbol: POM121C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015295907.1 | XP_015151393.1 | 8 - 9 | 1195 | 95 | |
| XM_015295908.1 | XP_015151394.1 | 8 - 9 | 1192 | 95 | ||
| GenBank mRNA | CR387663.1 | - | 4 - 5 | 461 | 95 | |
More Information
Set Membership: |
|

POM121C
Genomic Map