Assay Details
Gene Symbol: CZH5ORF51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_424778.4 | XP_424778.2 | 4 - 5 | 685 | 102 | |
GenBank mRNA | CR387715.1 | - | 4 - 5 | 483 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
