Assay Details
| Gene Symbol: CLIC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004935921.3 | XP_004935978.1 | 6 | 8661 | 99 | |
| XM_420060.5 | XP_420060.2 | 6 | 8920 | 99 | ||
| GenBank mRNA | CR388731.1 | - | 1 | 515 | 99 | |
More Information
Set Membership: |
|

CLIC5
Genomic Map