Assay Details
| Gene Symbol: DIMT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_424746.6 | XP_424746.3 | 8 - 9 | 706 | 83 | |
| GenBank mRNA | CR389591.1 | - | 3 - 4 | 276 | 83 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DIMT1| Gene Symbol: DIMT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_424746.6 | XP_424746.3 | 8 - 9 | 706 | 83 | |
| GenBank mRNA | CR389591.1 | - | 3 - 4 | 276 | 83 | |
Set Membership: |
|