Assay Details
Gene Symbol: SCRN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015299484.2 | XP_015154970.2 | 4 - 5 | 820 | 67 | |
XM_015299485.2 | XP_015154971.1 | 4 - 5 | 613 | 67 | ||
XM_025144122.1 | XP_024999890.1 | 4 - 5 | 1565 | 67 | ||
GenBank mRNA | CR389943.1 | - | 3 - 4 | 406 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
