Assay Details
| Gene Symbol: WSB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_415313.5 | XP_415313.3 | 8 - 9 | 1174 | 72 | |
| GenBank mRNA | CR390046.1 | - | 4 - 5 | 445 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

WSB2| Gene Symbol: WSB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_415313.5 | XP_415313.3 | 8 - 9 | 1174 | 72 | |
| GenBank mRNA | CR390046.1 | - | 4 - 5 | 445 | 72 | |
Set Membership: |
|