Assay Details
Gene Symbol: BHLHA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001235100.5 | XP_001235101.1 | 1 | 1007 | 93 | |
GenBank mRNA | CR390392.1 | - | 1 | 913 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
