Assay Details
Gene Symbol: LOC107050456 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025145463.1 | XP_025001231.1 | 11 - 12 | 1397 | 95 | |
GenBank mRNA | AB090806.1 | - | 5 - 6 | 493 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
