Assay Details
| Gene Symbol: CHDSD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003640648.4 | XP_003640696.1 | 3 - 4 | 524 | 97 | |
| XM_015281129.2 | XP_015136615.1 | 3 - 4 | 524 | 97 | ||
| GenBank mRNA | AB284370.1 | - | 2 - 3 | 432 | 97 | |
| BX929724.2 | - | 515 | 97 | |||
More Information
Set Membership: |
|

CHDSD
Genomic Map