Assay Details
Gene Symbol: RSPO4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004947141.3 | XP_004947198.2 | 2 - 3 | 658 | 96 | |
XM_004947142.3 | XP_004947199.1 | 2 - 3 | 1025 | 96 | ||
XM_004947143.3 | XP_004947200.2 | 2 - 3 | 633 | 96 | ||
XM_425713.6 | XP_425713.5 | 2 - 3 | 613 | 96 | ||
GenBank mRNA | AB690565.1 | - | 2 - 3 | 574 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
