Assay Details
| Gene Symbol: RALA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004939564.3 | XP_004939621.1 | 5 - 6 | 614 | 128 | |
| XM_015281637.2 | XP_015137123.1 | 3 - 4 | 502 | 128 | ||
| XM_025147578.1 | XP_025003346.1 | 7 - 8 | 4457 | 128 | ||
| XM_025147579.1 | XP_025003347.1 | 8 - 9 | 4544 | 128 | ||
| XM_025147580.1 | XP_025003348.1 | 4 - 5 | 482 | 128 | ||
| XM_418863.6 | XP_418863.1 | 3 - 4 | 399 | 128 | ||
| GenBank mRNA | CR405771.1 | - | 3 - 4 | 427 | 128 | |
More Information
Set Membership: |
|

RALA
Genomic Map