Assay Details
| Gene Symbol: TEKT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015297799.2 | XP_015153285.1 | 1 - 2 | 274 | 127 | |
| XM_015297800.2 | XP_015153286.1 | 2 - 3 | 312 | 127 | ||
| XM_417774.6 | XP_417774.3 | 2 - 3 | 5922 | 127 | ||
| GenBank mRNA | CR405823.1 | - | 1 - 2 | 342 | 127 | |
More Information
Set Membership: |
|

TEKT2
Genomic Map