Assay Details
| Gene Symbol: ZFAND4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015288445.2 | XP_015143931.1 | 3 - 4 | 385 | 84 | |
| XM_015288446.2 | XP_015143932.1 | 3 - 4 | 385 | 84 | ||
| XM_015288447.2 | XP_015143933.1 | 3 - 4 | 385 | 84 | ||
| XM_025151665.1 | XP_025007433.1 | 3 - 4 | 450 | 84 | ||
| XM_025151666.1 | XP_025007434.1 | 3 - 4 | 385 | 84 | ||
| XM_025151667.1 | XP_025007435.1 | 3 - 4 | 385 | 84 | ||
| GenBank mRNA | CR406033.1 | - | 2 - 3 | 317 | 84 | |
More Information
Set Membership: |
|

ZFAND4
Genomic Map