Assay Details
Gene Symbol: PAK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004936995.3 | XP_004937052.1 | 21 | 7077 | 83 | |
XM_025153752.1 | XP_025009520.1 | 22 | 7268 | 83 | ||
XM_025153753.1 | XP_025009521.1 | 20 | 6865 | 83 | ||
XM_422671.5 | XP_422671.2 | 20 | 6963 | 83 | ||
GenBank mRNA | CR406103.1 | - | 1 | 819 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
