Assay Details
| Gene Symbol: RAB18L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BX929919.2 | - | 3 - 4 | 467 | 90 | |
| CR405824.1 | - | 3 - 4 | 350 | 90 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RAB18L| Gene Symbol: RAB18L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BX929919.2 | - | 3 - 4 | 467 | 90 | |
| CR405824.1 | - | 3 - 4 | 350 | 90 | ||
Set Membership: |
|