Assay Details
| Gene Symbol: RSPH10B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003642138.4 | XP_003642186.2 | 18 - 19 | 3127 | 98 | |
| XM_004945097.3 | XP_004945154.1 | 18 - 19 | 3127 | 98 | ||
| XM_004945098.3 | XP_004945155.1 | 18 - 19 | 3127 | 98 | ||
| XM_004945099.3 | XP_004945156.1 | 17 - 18 | 2953 | 98 | ||
| XM_004945100.3 | XP_004945157.1 | 17 - 18 | 2950 | 98 | ||
| XM_004945101.3 | XP_004945158.1 | 17 - 18 | 2950 | 98 | ||
| GenBank mRNA | BX929963.1 | - | 7 - 8 | 872 | 98 | |
| BX934420.2 | - | 4 - 5 | 527 | 98 | ||
More Information
Set Membership: |
|

RSPH10B
Genomic Map