Assay Details
| Gene Symbol: PITPNC1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003642161.2 | XP_003642209.1 | 6 - 7 | 528 | 76 | |
| XM_004945414.3 | XP_004945471.1 | 6 - 7 | 528 | 76 | ||
| XM_015294715.2 | XP_015150201.1 | 5 - 6 | 438 | 76 | ||
| GenBank mRNA | BX930020.1 | - | 5 - 6 | 385 | 76 | |
More Information
Set Membership: |
|

PITPNC1L
Genomic Map