Assay Details
| Gene Symbol: LOC100857622 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015274577.2 | XP_015130063.1 | 3 - 4 | 423 | 66 | |
| GenBank mRNA | BX930073.2 | - | 3 - 4 | 412 | 66 | |
| BX931946.2 | - | 3 - 4 | 414 | 66 | ||
| BX950608.1 | - | 3 - 4 | 445 | 66 | ||
More Information
Set Membership: |
|

LOC100857622
Genomic Map