Assay Details
| Gene Symbol: LOC426355 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BX930207.1 | - | 1 | 617 | 107 | |
| BX931869.1 | - | 1 | 617 | 107 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC426355| Gene Symbol: LOC426355 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BX930207.1 | - | 1 | 617 | 107 | |
| BX931869.1 | - | 1 | 617 | 107 | ||
Set Membership: |
|