Assay Details
| Gene Symbol: ANAPC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_415533.6 | XP_415533.2 | 2 - 3 | 869 | 54 | |
| GenBank mRNA | CR407060.1 | - | 586 | 54 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ANAPC2| Gene Symbol: ANAPC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_415533.6 | XP_415533.2 | 2 - 3 | 869 | 54 | |
| GenBank mRNA | CR407060.1 | - | 586 | 54 | ||
Set Membership: |
|