Assay Details
| Gene Symbol: HOGA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001233351.5 | XP_001233352.2 | 5 - 6 | 736 | 66 | |
| GenBank mRNA | CR407189.1 | - | 3 - 4 | 268 | 66 | |
More Information
Set Membership: |
|

HOGA1
Genomic Map