Assay Details
Gene Symbol: PAK6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004941288.2 | XP_004941345.1 | 13 | 4060 | 115 | |
XM_004941289.2 | XP_004941346.1 | 12 | 3899 | 115 | ||
XM_004941290.3 | XP_004941347.1 | 10 | 3061 | 115 | ||
XM_004941291.3 | XP_004941348.1 | 9 | 3056 | 115 | ||
XM_004941292.3 | XP_004941349.1 | 9 | 3488 | 115 | ||
XM_015286277.1 | XP_015141763.1 | 13 | 4227 | 115 | ||
XM_015286278.2 | XP_015141764.1 | 12 | 5204 | 115 | ||
XM_015286279.1 | XP_015141765.1 | 11 | 3706 | 115 | ||
GenBank mRNA | CR523306.1 | - | 1 | 418 | 115 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
