Assay Details
Gene Symbol: TMEM35B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003642569.4 | XP_003642617.3 | 1 - 2 | 129 | 76 | |
GenBank mRNA | CR524040.1 | - | 1 - 2 | 119 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
