Assay Details
| Gene Symbol: RNF128 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004940850.3 | XP_004940907.1 | 7 | 5932 | 87 | |
| XM_420351.6 | XP_420351.2 | 7 | 2628 | 87 | ||
| GenBank mRNA | CR524065.1 | - | 1 | 921 | 87 | |
More Information
Set Membership: |
|

RNF128
Genomic Map