Assay Details
Gene Symbol: ZNF740 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015273414.2 | XP_015128900.1 | 5 - 6 | 639 | 60 | |
XM_015273415.2 | XP_015128901.1 | 5 - 6 | 667 | 60 | ||
XM_015273416.2 | XP_015128902.1 | 5 - 6 | 804 | 60 | ||
XM_015273417.2 | XP_015128903.1 | 5 - 6 | 606 | 60 | ||
GenBank mRNA | CR524262.1 | - | 4 - 5 | 436 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
