Assay Details
| Gene Symbol: FAM84B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004940033.3 | XP_004940090.1 | 2 | 5871 | 134 | |
| XM_015283142.2 | XP_015138628.1 | 2 | 5980 | 134 | ||
| XM_015283143.2 | XP_015138629.1 | 2 | 5680 | 134 | ||
| XM_025148158.1 | XP_025003926.1 | 2 | 5687 | 134 | ||
| GenBank mRNA | CR524351.1 | - | 1 | 814 | 134 | |
More Information
Set Membership: |
|

FAM84B
Genomic Map