Assay Details
| Gene Symbol: PRDX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_426543.5 | XP_426543.4 | 5 - 6 | 655 | 112 | |
| GenBank mRNA | CR733314.1 | - | 3 - 4 | 380 | 112 | |
More Information
Set Membership: |
|

PRDX3
Genomic Map