Assay Details
| Gene Symbol: GHRHR-LR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_425958.6 | XP_425958.3 | 1 - 2 | 515 | 80 | |
| GenBank mRNA | EF209053.1 | - | 1 - 2 | 67 | 80 | |
More Information
Set Membership: |
|

GHRHR-LR
Genomic Map