Assay Details
Gene Symbol: UPF3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025146692.1 | XP_025002460.1 | 4 - 5 | 915 | 143 | |
XM_025146693.1 | XP_025002461.1 | 4 - 5 | 915 | 143 | ||
XM_025146694.1 | XP_025002462.1 | 3 - 4 | 337 | 143 | ||
XM_025146695.1 | XP_025002463.1 | 3 - 4 | 337 | 143 | ||
XM_025146696.1 | XP_025002464.1 | 3 - 4 | 589 | 143 | ||
XM_416933.6 | XP_416933.2 | 4 - 5 | 907 | 143 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
