Assay Details
Gene Symbol: C1QTNF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004945218.3 | XP_004945275.3 | 2 - 3 | 631 | 97 | |
XM_425240.5 | XP_425240.3 | 2 - 3 | 1181 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
