Assay Details
| Gene Symbol: DLC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004936009.3 | XP_004936066.1 | 14 | 4850 | 122 | |
| XM_004936011.3 | XP_004936068.1 | 14 | 4579 | 122 | ||
| XM_015285550.2 | XP_015141036.1 | 20 | 6393 | 122 | ||
| XM_015285551.2 | XP_015141037.1 | 18 | 6226 | 122 | ||
| XM_025149937.1 | XP_025005705.1 | 18 | 6683 | 122 | ||
| XM_420693.6 | XP_420693.3 | 21 | 6319 | 122 | ||
More Information
Set Membership: |
|

DLC1
Genomic Map