Assay Details
Gene Symbol: WHAMM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025153945.1 | XP_025009713.1 | 7 - 8 | 1540 | 121 | |
XM_025153946.1 | XP_025009714.1 | 8 - 9 | 1233 | 121 | ||
XM_413835.6 | XP_413835.2 | 7 - 8 | 1540 | 121 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
