Assay Details
Gene Symbol: CFAP100 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015293360.2 | XP_015148846.1 | 14 - 15 | 1805 | 60 | |
XM_425162.6 | XP_425162.4 | 16 - 17 | 1985 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
