Assay Details
Gene Symbol: PQLC2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004943545.3 | XP_004943602.1 | 7 - 8 | 1045 | 106 | |
XM_004943548.3 | XP_004943605.1 | 5 - 6 | 1334 | 106 | ||
XM_015291787.2 | XP_015147273.1 | 7 - 8 | 1045 | 106 | ||
XM_015291788.2 | XP_015147274.1 | 7 - 8 | 783 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
