Assay Details
Gene Symbol: PQLC2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004943545.3 | XP_004943602.1 | 8 - 9 | 1230 | 116 | |
XM_004943548.3 | XP_004943605.1 | 6 - 7 | 1519 | 116 | ||
XM_015291788.2 | XP_015147274.1 | 8 - 9 | 968 | 116 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
