Assay Details
| Gene Symbol: ADAP1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015288469.2 | XP_015143955.1 | 2 - 3 | 937 | 74 | |
| XM_015288479.2 | XP_015143965.1 | 2 - 3 | 920 | 74 | ||
More Information
Set Membership: |
|

ADAP1L
Genomic Map