Assay Details
Gene Symbol: STN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015288718.2 | XP_015144204.1 | 8 - 9 | 920 | 77 | |
XM_015288719.2 | XP_015144205.1 | 9 - 10 | 1039 | 77 | ||
XM_015288721.2 | XP_015144207.1 | 8 - 9 | 1150 | 77 | ||
XM_025151943.1 | XP_025007711.1 | 8 - 9 | 1059 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
