Assay Details
| Gene Symbol: LGALS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015288411.2 | XP_015143897.1 | 2 - 3 | 196 | 104 | |
| XM_025155386.1 | XP_025011154.1 | 3 - 4 | 365 | 104 | ||
| GenBank mRNA | BX930674.1 | - | 3 - 4 | 379 | 104 | |
| CR352551.1 | - | 4 - 5 | 434 | 104 | ||
More Information
Set Membership: |
|

LGALS2
Genomic Map