Assay Details
Gene Symbol: C22orf39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015275693.2 | XP_015131179.1 | 2 - 3 | 265 | 92 | |
GenBank mRNA | BX930721.2 | - | 2 - 3 | 298 | 92 | |
CR387269.1 | - | 2 - 3 | 293 | 92 | ||
CR407476.1 | - | 2 - 3 | 301 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
