Assay Details
| Gene Symbol: AK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004947852.3 | XP_004947909.1 | 2 - 3 | 254 | 81 | |
| XM_425786.6 | XP_425786.2 | 2 - 3 | 273 | 81 | ||
| GenBank mRNA | BX930820.2 | - | 1 - 2 | 37 | 81 | |
| BX933252.1 | - | 1 - 2 | 73 | 81 | ||
| BX934483.2 | - | 1 - 2 | 100 | 81 | ||
| BX936084.2 | - | 2 - 3 | 241 | 81 | ||
More Information
Set Membership: |
|

AK2
Genomic Map