Assay Details
Gene Symbol: C4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004938740.3 | XP_004938797.1 | 36 - 37 | 4675 | 60 | |
XM_025146825.1 | XP_025002593.1 | 35 - 36 | 4435 | 60 | ||
GenBank mRNA | KX964631.1 | - | 35 - 36 | 4491 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
