Assay Details
| Gene Symbol: MRPL57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004938806.2 | XP_004938863.1 | 1 - 2 | 464 | 111 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004938806.2 transcripts.
Set Membership: |
|

MRPL57
Genomic Map