Assay Details
| Gene Symbol: MRPL42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004937627.3 | XP_004937684.1 | 5 - 6 | 579 | 86 | |
| XM_416141.6 | XP_416141.3 | 5 - 6 | 575 | 86 | ||
More Information
Set Membership: |
|

MRPL42
Genomic Map