Assay Details
| Gene Symbol: LOC424588 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015291283.2 | XP_015146769.1 | 16 | 1993 | 64 | |
| XM_015291285.2 | XP_015146771.1 | 16 | 1999 | 64 | ||
| XM_025153082.1 | XP_025008850.1 | 16 | 1772 | 64 | ||
| XM_422428.6 | XP_422428.3 | 15 | 1949 | 64 | ||
More Information
Set Membership: |
|

LOC424588
Genomic Map