Assay Details
Gene Symbol: C2H7orf31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025147436.1 | XP_025003204.1 | 8 - 9 | 894 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: C2H7orf31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025147436.1 | XP_025003204.1 | 8 - 9 | 894 | 72 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |