Assay Details
Gene Symbol: ADTRP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001234746.4 | XP_001234747.1 | 1 - 2 | 403 | 71 | |
XM_004939654.3 | XP_004939711.1 | 1 - 2 | 637 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
