Assay Details
| Gene Symbol: BRINP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004943288.3 | XP_004943345.1 | 8 | 2479 | 95 | |
| XM_426633.6 | XP_426633.2 | 8 | 2509 | 95 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

BRINP3| Gene Symbol: BRINP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004943288.3 | XP_004943345.1 | 8 | 2479 | 95 | |
| XM_426633.6 | XP_426633.2 | 8 | 2509 | 95 | ||
Set Membership: |
|