Assay Details
| Gene Symbol: C2H8ORF34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015282758.2 | XP_015138244.2 | 9 - 10 | 1406 | 74 | |
| XM_025147990.1 | XP_025003758.1 | 10 - 11 | 1536 | 74 | ||
More Information
Set Membership: |
|

C2H8ORF34
Genomic Map