Assay Details
| Gene Symbol: SLCO3A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_413876.5 | XP_413876.2 | 8 - 9 | 1677 | 61 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLCO3A1| Gene Symbol: SLCO3A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_413876.5 | XP_413876.2 | 8 - 9 | 1677 | 61 | |
Set Membership: |
|