Assay Details
| Gene Symbol: PLA2G4EL6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015287113.2 | XP_015142599.1 | 16 - 17 | 1759 | 124 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PLA2G4EL6| Gene Symbol: PLA2G4EL6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015287113.2 | XP_015142599.1 | 16 - 17 | 1759 | 124 | |
Set Membership: |
|